Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242379.1 | NP_001229308.1 | 7, 8 | 996 | ||
NM_001242381.1 | NP_001229310.1 | 7, 8 | 993 | |||
NM_001242383.1 | NP_001229312.1 | 7, 8 | 993 | |||
NM_033552.3 | NP_291030.2 | 7, 8 | 996 | |||
GenBank mRNA | AB033759.1 | - | 7, 8 | 917 | ||
AK043017.1 | - | 7, 8 | 972 | |||
BC039226.1 | - | 7, 8 | 945 | 4480841 | ||
HM149278.1 | - | 7, 8 | 957 | |||
HQ831517.1 | - | 7, 8 | 933 | |||
JF500487.1 | - | 7, 8 | 1073 | |||
JF500489.1 | - | 7, 8 | 1073 | |||
JX254917.1 | - | 7, 8 | 963 |