Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053080.3 | NP_444310.3 | 1, 2 | 157 | ||
GenBank mRNA | AF152359.1 | - | 1, 2 | 112 | ||
AF246711.1 | - | 1, 2 | 105 | |||
AF253409.1 | - | 1, 2 | 144 | |||
AF280404.1 | - | 1, 2 | 109 | |||
AK147006.1 | - | 1, 2 | 158 | |||
BC058277.1 | - | 1, 2 | 101 | 6515355 |