Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox4i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053091.2 | NP_444321.1 | 3 | 258 | ||
GenBank mRNA | AF271382.1 | - | 3 | 258 | ||
BC049623.1 | - | 2 | 267 | 6532529 |