Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053114.2 | NP_444344.1 | 2 | 356 | ||
GenBank mRNA | AK050551.1 | - | 2 | 358 | ||
BC107362.1 | - | 1 | 257 | 40056131 | ||
BC107363.2 | - | 1 | 257 | 40056132 |