Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033652.5 | NP_387501.1 | 3, 4 | 721 | ||
GenBank mRNA | AF226662.1 | - | 3, 4 | 504 | ||
AK044944.1 | - | 3, 4 | 723 | |||
BC109167.2 | - | 3, 4 | 642 | 40048180 | ||
BC109168.2 | - | 3, 4 | 642 | 40048181 |