Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170786.2 | NP_740756.1 | 2 | 480 | ||
GenBank mRNA | AK041204.1 | - | 2 | 315 | ||
BC027539.1 | - | 2 | 479 | 1429801 |
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U05342.1 | - | 11 | 1310 |