Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161782.1 | NP_001155254.1 | 2 | 695 | ||
NM_053084.2 | NP_444314.2 | 2 | 698 | |||
GenBank mRNA | AF347694.1 | - | 2 | 666 | ||
AK154028.1 | - | 2 | 663 | |||
AK166358.1 | - | 2 | 650 | |||
AX775777.1 | - | 2 | 678 | |||
BC034104.1 | - | 2 | 637 | 4502346 |