Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sep15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053102.2 | NP_444332.1 | 3 | 559 | ||
GenBank mRNA | AF288740.1 | - | 3 | 507 | ||
AK020419.1 | - | 3 | 273 | |||
AK159085.1 | - | 3 | 286 | |||
AK159353.1 | - | 3 | 268 | |||
BC010481.1 | - | 2 | 57 | |||
BC019792.1 | - | 3 | 275 | 5003585 |