Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053105.2 | NP_444335.2 | 1 | 1158 | ||
GenBank mRNA | AF252281.1 | - | 1 | 1087 | ||
AK031932.1 | - | 1 | 970 | |||
AK220397.1 | - | 1 | 462 | |||
BC094584.1 | - | 1 | 1158 | 30547502 |