Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Echs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053119.2 | NP_444349.1 | 3 | 340 | ||
GenBank mRNA | AK040391.1 | - | 3 | 341 | ||
AK044954.1 | - | 3 | 335 | |||
AK086762.1 | - | 3 | 333 | |||
AK088018.1 | - | 3 | 326 | |||
AK154752.1 | - | 2 | 134 | |||
AK167404.1 | - | 3 | 320 | |||
AK210204.1 | - | 2 | 152 | |||
BC002178.1 | - | 3 | 306 | |||
BC026722.1 | - | 2 | 695 | |||
BC033275.1 | - | 3 | 373 | |||
BC057971.1 | - | 2 | 64 | |||
BC072658.1 | - | 3 | 343 | 30357710 |