Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173372.1 | NP_001166843.1 | 9 | 418 | ||
NM_033648.1 | NP_387468.1 | 9 | 421 | |||
GenBank mRNA | AK018728.1 | - | 9 | 418 | ||
BC086918.1 | - | 10 | 682 | 6762406 |