Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003933.2 | NP_001003933.1 | 8 | 4472 | ||
NM_001003934.2 | NP_001003934.1 | 9 | 4529 | |||
NM_001271486.1 | NP_001258415.1 | 9 | 3566 | |||
NM_001271487.1 | NP_001258416.1 | 8 | 2405 | |||
NM_053076.3 | NP_444306.1 | 7 | 2348 | |||
GenBank mRNA | AF195940.1 | - | 7 | 2292 | ||
AK036892.1 | - | 7 | 2314 | |||
AK049845.1 | - | 7 | 2281 | |||
AK088670.1 | - | 7 | 2280 | |||
AK148792.1 | - | 7 | 2307 | |||
AK148947.1 | - | 7 | 2309 | |||
AK165683.1 | - | 7 | 2279 | |||
AY164700.1 | - | 7 | 2342 | |||
AY427822.1 | - | 8 | 4453 | |||
AY750849.1 | - | 9 | 4510 | |||
BC014697.1 | - | 7 | 2259 | |||
BC036717.1 | - | 7 | 2257 |