Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130868.2 | NP_001124340.1 | 9 | 1244 | ||
NM_001286384.1 | NP_001273313.1 | 10 | 1302 | |||
NM_053092.3 | NP_444322.1 | 8 | 1064 | |||
GenBank mRNA | AK089053.1 | - | 8 | 989 | ||
AK090203.1 | - | 8 | 989 | |||
AK153236.1 | - | 8 | 987 | |||
AK167693.1 | - | 8 | 989 | |||
BC027356.1 | - | 10 | 1214 | 4950333 | ||
BC035324.1 | - | 9 | 1156 | 2811286 | ||
BC036289.1 | - | 8 | 1033 | 3157706 |