Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053171.2 | NP_444401.2 | 2 | 530 | ||
GenBank mRNA | AK038679.1 | - | 2 | 511 | ||
AK048698.1 | - | 2 | 513 | |||
AK048713.1 | - | 2 | 732 | |||
AK134585.1 | - | 2 | 531 | |||
AY017475.1 | - | 2 | 427 |