Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197321.1 | NP_001184250.1 | 5 | 572 | ||
NM_001197322.1 | NP_001184251.1 | 2 | 176 | |||
NM_053202.2 | NP_444432.1 | 6 | 724 | |||
GenBank mRNA | AB221601.1 | - | 3 | 413 | ||
AB221602.1 | - | 3 | 413 | |||
AB594192.1 | - | 7 | 1129 | |||
AF339103.1 | - | 3 | 413 | |||
AF339104.1 | - | 3 | 413 | |||
AK077062.1 | - | 2 | 71 | |||
AK139831.1 | - | 2 | 62 | |||
AK144920.1 | - | 2 | 145 | |||
BC064764.1 | - | 5 | 558 | 30430449 |