Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700020M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006176.1 | - | 1 | 410 |
Gene Symbol: Gm15983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK140616.1 | - | 2 | 2135 |