Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000453.2 | NP_000444.1 | 5 | 937 | ||
XM_011528192.2 | XP_011526494.1 | 5 | 1516 | |||
XM_011528193.2 | XP_011526495.1 | 6 | 1019 | |||
XM_011528194.2 | XP_011526496.1 | 5 | 424 | |||
XM_017027158.1 | XP_016882647.1 | 6 | 1017 | |||
GenBank mRNA | BC105047.1 | - | 5 | 652 | 8144050 | |
BC105049.1 | - | 5 | 652 | 8144052 | ||
D87920.1 | - | 5 | 809 | |||
U66088.1 | - | 5 | 937 |