Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053147.3 | NP_444377.3 | 1 | 988 | ||
GenBank mRNA | AF326315.1 | - | 1 | 860 | ||
AK043492.1 | - | 1 | 989 | |||
AK143176.1 | - | 1 | 1469 | |||
AY013784.1 | - | 1 | 860 | |||
BC094239.1 | - | 2 | 461 | 30606144 |