Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053171.2 | NP_444401.2 | 22 | 3747 | ||
GenBank mRNA | AK081081.1 | - | 10 | 1747 | ||
AY017475.1 | - | 22 | 3644 |