Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000466.2 | NP_000457.1 | 2 | 291 | ||
NM_001282677.1 | NP_001269606.1 | 2 | 291 | |||
NM_001282678.1 | NP_001269607.1 | 2 | 291 | |||
GenBank mRNA | AB008112.1 | - | 2 | 262 | ||
AB052090.1 | - | 2 | 195 | |||
AB052091.1 | - | 2 | 195 | |||
AB052092.1 | - | 2 | 195 | |||
AB052093.1 | - | 2 | 195 | |||
AB052094.1 | - | 2 | 195 | |||
AF026086.1 | - | 2 | 255 | |||
AF030356.1 | - | 2 | 199 | |||
AK292955.1 | - | 2 | 196 | |||
AK293757.1 | - | 2 | 199 | |||
AK295686.1 | - | 2 | 201 | |||
BC035575.1 | - | 2 | 233 | 5498274 | ||
DA313282.1 | - | 2 | 292 | |||
DC370317.1 | - | 2 | 267 |