Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079873.1 | NP_001073342.1 | 4 | 582 | ||
NM_054054.2 | NP_473395.2 | 4 | 582 | |||
GenBank mRNA | AB208640.1 | - | 2 | 275 | ||
AF358660.1 | - | 4 | 617 | |||
AK077137.1 | - | 4 | 602 |