Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177773.1 | NP_001171244.1 | 6 | 1062 | ||
NM_001177774.1 | NP_001171245.1 | 5 | 971 | |||
NM_080428.3 | NP_536353.2 | 3 | 607 | |||
GenBank mRNA | AB221593.1 | - | 3 | 449 | ||
AF391192.1 | - | 3 | 607 | |||
AF427101.1 | - | 3 | 529 | |||
BC060620.1 | - | 6 | 1062 | |||
BC066093.1 | - | 5 | 971 | |||
BC085184.1 | - | 6 | 1161 | 30664813 | ||
BC131648.1 | - | 3 | 471 | 40110349 |