Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fezf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080433.3 | NP_536681.2 | 2 | 1066 | ||
GenBank mRNA | AB042399.1 | - | 1 | 783 | ||
AK019996.1 | - | 2 | 1030 | |||
BC055718.1 | - | 1 | 1393 | 6415359 |