Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080438.2 | NP_536686.2 | 2 | 174 | ||
GenBank mRNA | AF362764.1 | - | 2 | 95 | ||
AK038618.1 | - | 2 | 540 | |||
AY230204.1 | - | 2 | 113 |