Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253783.1 | NP_001240712.1 | 4, 5 | 325 | ||
NM_001253784.1 | NP_001240713.1 | 3, 4 | 303 | |||
NM_001281803.1 | NP_001268732.1 | 2, 3 | 307 | |||
NM_054046.5 | NP_473387.1 | 2, 3 | 216 | |||
GenBank mRNA | AK028726.1 | - | 3, 4 | 275 | ||
AK034028.1 | - | 182 | ||||
AK046271.1 | - | 2, 3 | 177 | |||
AK050405.1 | - | 3, 4 | 274 | |||
AK081295.1 | - | 2, 3 | 206 | |||
AK086174.1 | - | 2, 3 | 177 | |||
AK087778.1 | - | 2, 3 | 187 | |||
AK129019.1 | - | 2, 3 | 258 | |||
AK133979.1 | - | 2, 3 | 190 | |||
AK150334.1 | - | 2, 3 | 152 | |||
AK150994.1 | - | 2, 3 | 152 | |||
AK164147.1 | - | 2, 3 | 188 | |||
BC003306.1 | - | 4, 5 | 276 | 3154712 | ||
BY069015.1 | - | 3, 4 | 199 | |||
BY273027.1 | - | 2, 3 | 266 | |||
BY285979.1 | - | 2, 3 | 307 | |||
CX562217.1 | - | 2, 3 | 269 |