Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080443.2 | NP_536691.2 | 4 | 905 | ||
GenBank mRNA | AF398968.1 | - | 1 | 190 | ||
AK051017.1 | - | 5 | 893 | |||
BC062948.1 | - | 1 | 134 | 30532932 | ||
BC066824.1 | - | 3 | 407 | 30087227 | ||
BC085155.1 | - | 4 | 885 |