Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199122.1 | NP_001186051.1 | 9 | 2142 | ||
NM_078478.5 | NP_510963.1 | 9 | 2095 | |||
GenBank mRNA | AF412297.1 | - | 9 | 2040 | ||
AK088883.1 | - | 1 | 68 | |||
AK093887.1 | - | 9 | 2071 | |||
AK134592.1 | - | 9 | 2025 | |||
AK146433.1 | - | 9 | 2002 | |||
AK146853.1 | - | 9 | 2058 | |||
AK149244.1 | - | 1 | 589 | |||
AK158904.1 | - | 9 | 2085 | |||
AK168678.1 | - | 9 | 2143 | |||
AK168800.1 | - | 9 | 2051 |