Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130457.2 | NP_569724.2 | 2 | 229 | ||
GenBank mRNA | AF333770.1 | - | 2 | 194 | ||
AK042607.1 | - | 2 | 218 | |||
AK053611.1 | - | 2 | 221 | |||
AK081891.1 | - | 2 | 222 | |||
AK142816.1 | - | 2 | 225 | |||
AK173255.1 | - | 2 | 204 | |||
BC034628.1 | - | 2 | 229 | 5369648 |