Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080556.3 | NP_542123.3 | 4 | 587 | ||
GenBank mRNA | AK010325.1 | - | 4 | 544 | ||
AK030340.1 | - | 4 | 517 | |||
AK048012.1 | - | 4 | 506 | |||
AK050338.1 | - | 4 | 534 | |||
AK075720.1 | - | 4 | 587 | |||
AK088905.1 | - | 4 | 559 | |||
AK206314.1 | - | 1 | 66 | |||
BC003862.1 | - | 4 | 526 | 3488326 |