Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146324.1 | NP_001139796.1 | 2 | 395 | ||
NM_080634.4 | NP_542365.3 | 2 | 395 | |||
GenBank mRNA | AF393747.1 | - | 2 | 384 | ||
AF393780.1 | - | 2 | 387 | |||
AK034377.1 | - | 2 | 366 | |||
AK036663.1 | - | 2 | 403 | |||
AK037829.1 | - | 1 | 43 | |||
AK049204.1 | - | 2 | 388 | |||
AK079896.1 | - | 2 | 387 | |||
AK086167.1 | - | 2 | 384 | |||
AK155497.1 | - | 2 | 362 | |||
AK157953.1 | - | 2 | 380 | |||
AK166194.1 | - | 2 | 380 | |||
AK169855.1 | - | 2 | 366 | |||
AK200638.1 | - | 1 | 78 | |||
BC058235.1 | - | 2 | 369 | 6307400 |