Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000562.2 | NP_000553.1 | 1, 2 | 209 | ||
XM_011542079.2 | XP_011540381.1 | 1, 2 | 183 | |||
XM_017002234.1 | XP_016857723.1 | 1, 2 | 183 | |||
GenBank mRNA | AK309596.1 | - | 1, 2 | 183 | ||
BC020702.1 | - | 1, 2 | 182 | |||
BC132911.1 | - | 1, 2 | 101 | 40146933 | ||
BC132913.1 | - | 1, 2 | 101 | 40146935 | ||
M16974.1 | - | 1, 2 | 214 |