Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130860.3 | NP_570930.1 | 3 | 330 | ||
GenBank mRNA | AF327431.1 | - | 3 | 202 | ||
AK089276.1 | - | 3 | 169 | |||
AK142397.1 | - | 1 | 365 | |||
AK143217.1 | - | 3 | 331 | |||
AK144607.1 | - | 3 | 311 | |||
AK155733.1 | - | 2 | 541 | |||
AK157340.1 | - | 3 | 302 | |||
AK206169.1 | - | 3 | 224 | |||
BC003901.1 | - | 3 | 275 | 3601310 |