Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap19-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130870.1 | NP_570940.1 | 1 | 485 | ||
GenBank mRNA | AF345291.1 | - | 1 | 485 | ||
AK137740.1 | - | 1 | 181 | |||
BC115787.1 | - | 1 | 482 | 40105094 | ||
BC115815.1 | - | 1 | 485 | 40105095 |