Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080595.2 | NP_542162.1 | 15 | 1401 | ||
GenBank mRNA | AJ416093.1 | - | 15 | 1401 | ||
AK153702.1 | - | 17 | 1579 | |||
BC012248.1 | - | 6 | 503 | |||
BC021849.1 | - | 5 | 405 | |||
BC130023.1 | - | 15 | 1406 | 40140754 |