Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080795.4 | NP_542985.4 | 10 | 3135 | ||
GenBank mRNA | AF401681.1 | - | 10 | 3092 | ||
AK036218.1 | - | 10 | 3136 | |||
AK047053.1 | - | 10 | 3122 | |||
AK088210.1 | - | 1 | 230 | |||
AK140320.1 | - | 10 | 3070 | |||
AK170146.1 | - | 10 | 3103 | |||
BC090665.1 | - | 10 | 3101 | 30686384 | ||
BC117937.1 | - | 10 | 3088 | 40097802 | ||
BC117938.1 | - | 10 | 3089 | 40097810 |