Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133485.2 | NP_597844.1 | 1 | 729 | ||
GenBank mRNA | AF407167.2 | - | 1 | 826 | ||
AK042973.1 | - | 1 | 678 | |||
AK047607.1 | - | 1 | 738 | |||
AK085997.1 | - | 1 | 767 | |||
BC055717.1 | - | 1 | 336 | 6415732 |
Gene Symbol: Gm14057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013448.1 | - | 1 | 81 | ||
BC057040.1 | - | 1906 |