Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085409.1 | NP_001078878.1 | 5 | 1505 | ||
NM_133186.3 | NP_573449.2 | 4 | 1132 | |||
GenBank mRNA | AK003108.1 | - | 1106 | |||
AK028961.1 | - | 4 | 1109 | |||
AK078769.1 | - | 4 | 1526 | |||
AK167028.1 | - | 5 | 1505 | |||
AK171237.1 | - | 4 | 1137 | |||
AK217925.1 | - | 2 | 212 | |||
AY029586.1 | - | 4 | 1120 | |||
AY214462.1 | - | 1498 | ||||
BC027251.1 | - | 4 | 1115 | |||
BC037435.1 | - | 4 | 1133 | 2651584 |