Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prima1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133364.2 | NP_579942.2 | 5 | 485 | ||
GenBank mRNA | AK021000.1 | - | 3 | 431 | ||
AK039705.1 | - | 3 | 349 | |||
AK140034.1 | - | 6 | 748 | |||
AY043275.1 | - | 5 | 486 | |||
AY225515.1 | - | 6 | 657 | |||
BC134395.1 | - | 5 | 502 | |||
BC145454.1 | - | 4 | 329 | |||
BC171927.1 | - | 5 | 461 | |||
BC171928.1 | - | 5 | 500 | |||
BC171929.1 | - | 5 | 456 |