Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB052336.1 | - | 1 | 149 | ||
AB052337.1 | - | 1 | 149 | |||
AB052338.1 | - | 1 | 149 | |||
AF475079.1 | - | 1 | 149 | |||
AF475080.1 | - | 1 | 149 | |||
BC141258.1 | - | 1 | 208 | 9055870 | ||
BC145600.1 | - | 1 | 208 | 9054166 |
Gene Symbol: 6530402F18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079298.1 | - | 2 | 1508 | ||
AK154396.1 | - | 2 | 1445 | |||
AK220426.1 | - | 792 | ||||
BC066084.1 | - | 2 | 1516 |