Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133758.3 | NP_598519.2 | 17 | 2206 | ||
NM_177249.3 | NP_796223.2 | 18 | 2266 | |||
GenBank mRNA | AK030909.1 | - | 2 | 157 | ||
AK131138.1 | - | 8 | 883 | |||
BC024117.1 | - | 1 | 32 | |||
BC108425.1 | - | 18 | 2207 | 30062566 |