Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133922.3 | NP_598683.3 | 4 | 623 | ||
GenBank mRNA | AB041543.1 | - | 3 | 456 | ||
AK040525.1 | - | 4 | 625 | |||
AK128931.1 | - | 4 | 577 | |||
AK162934.1 | - | 4 | 331 | |||
AK173281.1 | - | 4 | 2327 | |||
AK191169.1 | - | 1 | 30 | |||
AK203900.1 | - | 1 | 30 | |||
BC066816.1 | - | 4 | 543 | 30084911 |