Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgdia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133796.7 | NP_598557.3 | 6 | 591 | ||
GenBank mRNA | AK075656.1 | - | 5 | 458 | ||
AK077343.1 | - | 6 | 751 | |||
AK143516.1 | - | 6 | 575 | |||
AK147842.1 | - | 550 | ||||
AK156819.1 | - | 536 | ||||
AK165241.1 | - | 6 | 573 | |||
AK170553.1 | - | 6 | 558 | |||
AK170718.1 | - | 6 | 575 | |||
AK172140.1 | - | 536 | ||||
AK201731.1 | - | 2 | 99 | |||
AK201760.1 | - | 2 | 99 | |||
AK205658.1 | - | 2 | 99 | |||
AK217147.1 | - | 2 | 92 | |||
BC004732.1 | - | 6 | 533 | 3583026 | ||
BC086755.1 | - | 6 | 515 | 6414768 |