Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133807.1 | NP_598568.1 | 7 | 1412 | ||
GenBank mRNA | AK053450.1 | - | 7 | 1439 | ||
AK150916.1 | - | 2 | 724 | |||
AK159374.1 | - | 7 | 1468 | |||
AK159552.1 | - | 7 | 1462 | |||
AK166576.1 | - | 7 | 1461 | |||
AK167604.1 | - | 7 | 1403 | |||
AK169250.1 | - | 5 | 1024 | |||
AK169390.1 | - | 7 | 1411 | |||
AK212896.1 | - | 1 | 291 | |||
AK213336.1 | - | 1 | 222 | |||
BC006877.1 | - | 7 | 1412 | 2647513 | ||
BC066172.1 | - | 7 | 1406 | 30429192 |