Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133810.3 | NP_598571.2 | 4 | 624 | ||
GenBank mRNA | AK029551.1 | - | 4 | 565 | ||
AK035693.1 | - | 4 | 592 | |||
AK076932.1 | - | 4 | 591 | |||
AK083332.1 | - | 4 | 593 | |||
AK153108.1 | - | 4 | 568 | |||
AK165291.1 | - | 4 | 564 | |||
AY092028.1 | - | 4 | 577 | |||
BC006579.1 | - | 4 | 579 | 3586767 |