Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133879.2 | NP_598640.2 | 3 | 886 | ||
GenBank mRNA | AK027912.1 | - | 2 | 1255 | ||
AM161454.1 | - | 3 | 834 | |||
BC005515.1 | - | 3 | 897 | 3493611 | ||
CX202255.1 | - | 1 | 63 |