Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133922.3 | NP_598683.3 | 10, 11 | 1692 | ||
GenBank mRNA | AB041543.1 | - | 8, 9 | 1324 | ||
AK029235.1 | - | 1, 2 | 138 | |||
AK040525.1 | - | 10, 11 | 1694 | |||
AK128931.1 | - | 10, 11 | 1643 | |||
AK162934.1 | - | 10, 11 | 1400 | |||
AK173281.1 | - | 10, 11 | 3396 | |||
AK197252.1 | - | 1, 2 | 40 | |||
BC066816.1 | - | 10, 11 | 1612 | 30084911 |