Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134034.2 | NP_598795.2 | 2, 3 | 605 | ||
GenBank mRNA | AK037253.1 | - | 2, 3 | 537 | ||
AK041513.1 | - | 2, 3 | 550 | |||
AK044742.1 | - | 2, 3 | 546 | |||
AK090014.1 | - | 2, 3 | 581 | |||
AK129344.1 | - | 2, 3 | 516 | |||
AK132660.1 | - | 2, 3 | 127 | |||
AK165940.1 | - | 2, 3 | 529 | |||
AK166627.1 | - | 2, 3 | 531 | |||
BC006870.1 | - | 2, 3 | 516 | 3595439 | ||
CD350450.1 | - | 2, 3 | 605 |