Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133932.2 | NP_598693.1 | 5 | 901 | ||
GenBank mRNA | AF383154.1 | - | 5 | 720 | ||
AK078906.1 | - | 5 | 1221 | |||
AK211996.1 | - | 2 | 230 | |||
BC022707.1 | - | 3 | 328 | 4486171 | ||
BC026630.1 | - | 5 | 901 | 4983657 | ||
BC032195.1 | - | 6 | 765 | 5323331 |