Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftsj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290430.1 | NP_001277359.1 | 11, 12 | 1222 | ||
NM_133991.2 | NP_598752.2 | 11, 12 | 1228 | |||
GenBank mRNA | AK036313.1 | - | 11, 12 | 1186 | ||
AK150251.1 | - | 8, 9 | 1316 | |||
BC011144.1 | - | 10, 11 | 1084 | 4194843 | ||
BC034880.1 | - | 11, 12 | 1092 | 3468633 |