Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134050.4 | NP_598811.3 | 3, 4 | 461 | ||
GenBank mRNA | AB232610.1 | - | 3, 4 | 254 | ||
AK158637.1 | - | 3, 4 | 383 | |||
AK158768.1 | - | 3, 4 | 415 | |||
BC013790.1 | - | 3, 4 | 407 | 2655151 | ||
BC027769.1 | - | 3, 4 | 461 | 5345297 |